Learn More
Thermo Scientific™ IMAGEN™ Herpes Simplex Virus Positive Control Slide
Validate your test procedures with Thermo Scientific™ IMAGEN™ Herpes Simplex Virus Positive Control Slide
Brand: Thermo Scientific™ S611030-2
90.07 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Validate your test procedures with Thermo Scientific™ IMAGEN™ Herpes Simplex Virus Positive Control Slide. A Teflon-coated glass microscope slide with two separate 6mm diameter well areas containing acetone-fixed human fibroblast cells infected with either herpes simplex virus type 1 (HSV 1) or herpes simplex virus type 2 (HSV 2).
Sealed in foil pouch filled with nitrogen.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
Specifications
IMAGEN Herpes Simplex Virus Positive Control Slide | |
1 Slide |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6198974 | Certificate of Analysis | 20/11/2024 | S611030-2 |
6135732 | Certificate of Analysis | 30/10/2024 | S611030-2 |
3773426 | Certificate of Analysis | 05/04/2024 | S611030-2 |
3604289 | Certificate of Analysis | 02/06/2023 | S611030-2 |
3535154 | Certificate of Analysis | 09/12/2022 | S611030-2 |