Learn More
BRAND™ Impact-Resistant Polystyrene Laboratory Spatulas
Constructed of impact-resistant polystyrene. Available with double spatulate ends or a spatulate and spoon combination.
Brand:  BRAND™ 139817
28.50 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11813970
  • 29.30 EUR / Pack of 10
Estimated Shipment: 21-03-2025
Description
  • Impact-resistant polystyrene construction
  • Green in color
  • Supplied in packs of 10
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 Pk.
Green
Polystyrene
Spatula/spoon combination
Spatula
180 mm
Documents
One moment while we fetch your results.
Product Title
BRAND™ Impact-Resistant Polystyrene Laboratory Spatulas > Length: 180mm; spatula/spoon