Learn More
Terumo™ Insulin Syringe
Use these syringes for the aspiration of fluids or sampling blood or other body fluids. Terumo™ Insulin Syringes feature a unique double beveled design for greater patient comfort.
Brand:  Terumo™ 8MDSS01S2613E
Product Code. 17132714
  • 25.65 EUR / Pack of 100
Edge
Description
  • Ultra-sharp needles
  • Clear barrels
  • Bold, precise scale markings
  • Smooth plunger action
  • Tamper evident peel packs
  • Sterile, Individually wrapped
TRUSTED_SUSTAINABILITY
Specifications
Specifications
26 G
Terumo Insulin Syringe
1 mL
13 mm
100 Pack
Sterile, Individually Wrapped
Documents
One moment while we fetch your results.
Product Title
Terumo™ Insulin Syringe > Volume: 1 mL