missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 5 Rabbit anti-Human, Mouse, Rat, Clone: S04-0G6, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-15048-100UL
This item is not returnable.
View return policy
Description
Integrin beta 5 Monoclonal antibody specifically detects Integrin beta 5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Integrin beta 5 | |
Monoclonal | |
Unconjugated | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
Rabbit | |
Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
S04-0G6 | |
Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
FLJ26658, integrin beta-5, integrin, beta 5 | |
Recombinant protein of human Integrin beta 5 (Uniprot#: P18084) | |
100 μg | |
Cancer, Cellular Markers, Signal Transduction | |
3693 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction