missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 5 Rabbit anti-Human, Mouse, Rat, Clone: S04-0G6, Novus Biologicals™
Rabbit Monoclonal Antibody
285.09 EUR - 577.74 EUR
Specifications
Antigen | Integrin beta 5 |
---|---|
Clone | S04-0G6 |
Dilution | Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18331327
|
Novus Biologicals
NBP3-15048-25UL |
25 μg |
285.09 EUR
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18314944
|
Novus Biologicals
NBP3-15048-100UL |
100 μg |
577.74 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Integrin beta 5 Monoclonal antibody specifically detects Integrin beta 5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Integrin beta 5 | |
Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
FLJ26658, integrin beta-5, integrin, beta 5 | |
Recombinant protein of human Integrin beta 5 (Uniprot#: P18084) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S04-0G6 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer, Cellular Markers, Signal Transduction | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
3693 | |
IgG | |
Affinity-purified |