Learn More
3M™ E-A-R™ E-A-Rsoft Yellow Earplugs
Offers high attenuation and are made from a slow expanding, PU foam material. 3M E-A-R Polyurethane Earplug – 36dB.
Brand: 3M™ 7000052845
Description
- Made from a slow expanding, PU foam material
- Provides evenly distributed pressure, giving flexibility, and a good seal with optimum comfort.
- Bright, neon color makes it appealing to wearers
Specifications
Yellow Neon | |
Vinyl | |
36 dB | |
Ear Plug | |
Roll-down |
CE Approved | |
200/Pk. | |
Corded | |
Polyurethane |