Learn More
Invitrogen™ LR Clonase II for BaculoDirect™ Kits
Eukaryotic system for high-level protein expression in insect cells
Brand:  Invitrogen™ 11791023
307.04 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.
Product Code. 10317993
  • 404.00 EUR / 10 reactions
Explore more special offers
Description
Description
  • Combination of Gateway Technology with baculovirus expression makes BaculoDirect System fastest and easiest method for production of recombinant baculovirus
Cloning, Gateway Cloning, Gateway Recombination
Order Info
Shipping Conditions: Dry ice
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store LR Clonase™ mix at -80°C. All components are guaranteed stable for 6 months if stored properly.
LR Clonase
LR Clonase II
Dry Ice
10 reactions
BaculoDirect™, Clonase™, Gateway™
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ LR Clonase II for BaculoDirect™ Kits > 10 reactions
For Research Use Only. Not for use in diagnostic procedures.