Learn More
Invitrogen™ E-Gel™ Safe Imager™ E-Gel Real-Time Transilluminator
Watch migration of DNA in real-time in E-Gels containing SYBR™ Safe
Brand: Invitrogen™ G6500
Includes: One E-Gel Safe Imager Real Time Transilluminator unit, amber filter, and Safe Imager viewing glasses. Power cord not included. Requires E-Gel iBase Power System for operation.
Description
- Real-Time monitoring of DNA migration
- Improved safety for the user
- Enhanced cloning efficiency of retrieved DNA
- Real-Time Transilluminator designed for Safety and Cloning
- E-Geltrademark (TM) Safe Imager Real-Time Transilluminator is designed to fit perfectly beneath the E-Gel iBasetrademark (TM) Power System and is intended for use with E-Gels containing SYBRtrademark (TM) Safe
- Blue light emitted by the instrument is ideal for viewing DNA that is stained with SYBR Safe
- Unlike UV, blue light causes minimal damage to DNA, offering improved safety for the user and enhanced cloning efficiency of any DNA retrieved from gels
- Real-Time Transilluminator can be purchased as stand-alone item, in combination with E-Gel iBase Power System, or as part of Starter Kit that also includes an E-Gel iBase Power System, E-Gel CloneWelltrademark (TM) gels, and an E-Gel 96 High Range DNA Marker
- Self-contained Power System
- E-Gel iBase Power System is compatible with all low throughput E-Gels
- When running E-Gels containing SYBR Safe, you can safely watch DNA bands migrate through gel by using the E-Gel Safe Imager Real-Time Transilluminator together with E-Gel iBase Power System
- Use of SYBR Safe DNA Gel Stain and Safe Imager Transilluminator ensures maximum possible safety to user and gives improved cloning efficiency over ethidium bromide and UV
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Specifications
Real-Time Transilluminator | |
1 Each |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Manual: E-Gel iBase Power System E-Gel Safe Imager Real-time Transilluminator - Japanese
Version: Rev D 2007_06_19
File size: 1.3 MB
E-Gel® iBase™ Power System
Version: 20 September 2012
File size: 800.7 kB
Product Content Correction
Product Title
Invitrogen™ E-Gel™ Safe Imager™ E-Gel Real-Time Transilluminator > Real-Time Transilluminator