Learn More
Invitrogen™ pCR™8/GW/TOPO™ TA Cloning Kit with One Shot™ Mach1™-T1R E. coli and PureLink™ Kit
Delivers simple, 5-minute cloning and access to Gateway recombination cloning vector platform
Brand: Invitrogen™ K252002
719.58 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Includes: pCR8/GW/TOPO Vector, dNTPs, Salt Solution, Sterile Water, Gateway Forward and Reverse Sequencing Primers, Control Template and PCR Primers; TOP10 Chemically Competent E. coli Chemically Competent E. coli, S.O.C. Medium, and a pUC19 control plasmid. The PureLink Quick Plasmid Miniprep Kit contains Resuspension Buffer, RNase A, Lysis Buffer, Precipitation Buffer, Wash Buffer, TE Buffer, Spin Columns, and Wash and Recovery Tubes.
Description
- pCR™8 TOPO TA vector contains minimal MCS, so sequence more of insert and less of vector
- Patented att sites make pCR™8 vector is simplest way to create an entry clone, which facilitates easy access via Gateway recombination cloning to variety of expression vectors without tedious sub-cloning, saving valuable research time
- pCR™8/GW/TOPO TA Cloning Kit features fast growing E. coli that shortens cloning time
- 3'-T overhangs for direct ligation of Taq-amplified PCR products
- Sequencing primer sites within 55 bp of insertion site
- Spectinomycin resistance gene for robust selection in E. coli
- EcoR I sites flanking the PCR product insertion site for easy excision of inserts
Cloning,Gateway Cloning,PCR Cloning
Specifications
TOPO™ with Gateway™ | |
20 preps | |
TA Cloning Kit | |
pCR |
pCR™8/GW/TOPO™ Vector, dNTPs, Salt Solution, Sterile Water, Gateway Forward and Reverse Sequencing Primers, Control Template and PCR Primers; One Shot™ TOP10 Chemically Competent E. coli Chemically Competent E. coli, S.O.C. Medium, and a pUC19 control plasmid. The PureLink™ Quick Plasmid Miniprep Kit contains Resuspension Buffer, RNase A, Lysis Buffer, Precipitation Buffer, Wash Buffer, TE Buffer, Spin Columns, and Wash and Recovery Tubes. Store all components of the PureLink™ kit at room temperature. Store the One Shot™ E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored. | |
One Shot™ | |
20 Preps |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
pCR™8/GW/TOPO® TA Cloning® Kit
Version: 23 March 2012
File size: 476.9 kB
Product Content Correction
Product Title
Invitrogen™ pCR™8/GW/TOPO™ TA Cloning Kit with One Shot™ Mach1™-T1R E. coli and PureLink™ Kit > 20 preps