Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data - Temperature
For one free-selectable temperature value, Includestemperature distribution survey at Memmert to DIN 12880:2007-05
Brand:  Memmert™ IPP750ecoD00127
 View more versions of this product
Product Code. 17110265
  • 2175.00 EUR / Each
Explore more special offers
Description
Description
  • 27 measuring points (sizes 110 - 1060, 2200)
  • 26 measuring points (size 1400)
  • PQ checklist as support for validation by customer
Specifications
Specifications
IPP750eco
IQ/OQ Document with Device-specific Works Test Data
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ IQ/OQ Document with Device-Specific Works Test Data - Temperature > IPP750eco