missing translation for 'onlineSavingsMsg'
Learn More
Iron(II) Sulfate Heptahydrate, ACS Reagent Grade, Scharlab Product Code.: 11314204
compliance-icons

Iron(II) Sulfate Heptahydrate, ACS Reagent Grade, Scharlab

Product Code. 11314204
500 g
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy

Product Code. 11314204

Brand: Scharlab HI03510500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in analytical chemistry for determining chemical oxygen demand (COD).

  • Synonym: Iron vitriol
  • Molecular Formula: FeSO4 • 7H2O
  • Molar Mass: 278.02g/mol
  • CAS: 7782-63-0
  • EIN ECS No.: 231-753-5
  • Substances Not Precipitated by Ammonium Hydroxide: 0.05% max

Specifications

Melting Point >60°C (release of crystalline water)
CAS 7782-63-0
pH 3.0–4.0 (5%, water)
Quantity 500 g
Molecular Formula FeH14O11S
MDL Number MFCD00149719
Synonym iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
IUPAC Name λ²-iron(2+) heptahydrate sulfate
Molecular Weight (g/mol) 278.01
PubChem CID 62662
ChEBI CHEBI:75836
Formula Weight 278.02g/mol
Percent Purity 99.5% min (permanganometric)
Grade ACS Reagent
Identification Passes Test
Show More Show Less
compliance-icons
Product Identifier
  • Iron(II) sulfate heptahydrate, reagent grade, ACS, ISO, Reag .Ph Eur
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iron(II) Sulfate Heptahydrate, ACS Reagent Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.