Learn More
Isophorone diisocyanate, mixture of isomers, 98%, Thermo Scientific Chemicals
Product Code. 11323227
Click to view available options
Quantity:
5 g
100 g
500 g
Isophorone diisocyanate is useful for the preparation of polyurethane plastics. It is also used to make polyurethane coatings and in enamel coatings, which are resistant to abrasion and degradation from ultraviolet light. Further, it is used as coatings for automotive, flooring and roofing as well as paint applied to aircraft.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isophorone diisocyanate is useful for the preparation of polyurethane plastics. It is also used to make polyurethane coatings and in enamel coatings, which are resistant to abrasion and degradation from ultraviolet light. Further, it is used as coatings for automotive, flooring and roofing as well as paint applied to aircraft.
Solubility
Miscible with esters, ketones, ethers, and aromatic and aliphatic hydrocarbons.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and alcohols.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4098-71-9
Molecular Formula C12H18N2O2
Molecular Weight (g/mol) 222.29
MDL Number MFCD00064956
InChI Key NIMLQBUJDJZYEJ-UHFFFAOYNA-N
Synonym isophorone diisocyanate, ipdi, isophorone diamine diisocyanate, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethylcyclohexane, 3-isocyanatomethyl-3,5,5-trimethylcyclohexylisocyanate, cyclohexane, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethyl, 3-isocyanatomethyl-3,5,5-trimethylcyclohexyl isocyanate, ccris 6252, isocyanic acid, methylene 3,5,5-trimethyl-3,1-cyclohexylene ester, isophorone diisocyanate diisocyanates
PubChem CID 169132
ChEBI CHEBI:53214
IUPAC Name 5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane
SMILES CC1(C)CC(CC(C)(CN=C=O)C1)N=C=O
Specifications
Melting Point -60°C
Density 1.061
Boiling Point 153°C (10 mmHg)
Flash Point 163°C (325°F)
Refractive Index 1.484
Quantity 100 g
UN Number UN2290
Beilstein 2726467
Sensitivity Moisture sensitive
Solubility Information Miscible with esters,ketones,ethers,and aromatic and aliphatic hydrocarbons.
Formula Weight 222.29
Percent Purity 98%
Chemical Name or Material Isophorone diisocyanate, mixture of isomers