missing translation for 'onlineSavingsMsg'
Learn More

Isophorone diisocyanate, mixture of isomers, 98%, Thermo Scientific Chemicals

31.26 EUR - 109.55 EUR

Chemical Identifiers

CAS 4098-71-9
Molecular Formula C12H18N2O2
Molecular Weight (g/mol) 222.29
MDL Number MFCD00064956
InChI Key NIMLQBUJDJZYEJ-UHFFFAOYNA-N
Synonym isophorone diisocyanate, ipdi, isophorone diamine diisocyanate, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethylcyclohexane, 3-isocyanatomethyl-3,5,5-trimethylcyclohexylisocyanate, cyclohexane, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethyl, 3-isocyanatomethyl-3,5,5-trimethylcyclohexyl isocyanate, ccris 6252, isocyanic acid, methylene 3,5,5-trimethyl-3,1-cyclohexylene ester, isophorone diisocyanate diisocyanates
PubChem CID 169132
ChEBI CHEBI:53214
IUPAC Name 5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane
SMILES CC1(C)CC(CC(C)(CN=C=O)C1)N=C=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11313227
SDS View Documents
Thermo Scientific Alfa Aesar
L13759.06
5 g
31.26 EUR
5g
Please to purchase this item. Need a web account? Register with us today!
11323227
SDS View Documents
Thermo Scientific Alfa Aesar
L13759.22
100 g
42.31 EUR
100g
Please to purchase this item. Need a web account? Register with us today!
11333227
SDS View Documents
Thermo Scientific Alfa Aesar
L13759.36
500 g
109.55 EUR
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Isophorone diisocyanate is useful for the preparation of polyurethane plastics. It is also used to make polyurethane coatings and in enamel coatings, which are resistant to abrasion and degradation from ultraviolet light. Further, it is used as coatings for automotive, flooring and roofing as well as paint applied to aircraft.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isophorone diisocyanate is useful for the preparation of polyurethane plastics. It is also used to make polyurethane coatings and in enamel coatings, which are resistant to abrasion and degradation from ultraviolet light. Further, it is used as coatings for automotive, flooring and roofing as well as paint applied to aircraft.

Solubility
Miscible with esters, ketones, ethers, and aromatic and aliphatic hydrocarbons.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and alcohols.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H330-H334-H315-H319-H317-H335-H204
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Fire or projection hazard.

P260-P264b-P271-P272-P280-P284-P285-P302+P352-P304+P340-P305+P351+P338-P310-P312-P333+P313-P362-P501c

H315-H317-H319-H330-H334-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOPHORONE DIISOCYANATE

missing translation for 'einecsNumber' : 223-861-6

missing translation for 'rtecsNumber' : NQ9370000

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Isophorone diisocyanate, mixture of isomers, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.