Learn More
GPE Scientific™ J Young Vacuum/Schlenk Flask with O-Ring Tap and O-Ring Ball Joint Cup
GPE Scientific™ J Young Vacuum/Schlenk Flask with O-Ring Tap and O-Ring Ball Joint Cup is designed for use with Schlenk lines and a vacuum manifold and available in various capacities.
Brand: GPE Scientific™ CSR/50
This item is not returnable.
View return policy
Description
- Designed with a side arm for evacuating or filling with gases
- For connection to Schlenk lines
- Round-bottomed shape
- Includes O-ring tap
- Includes O-ring ball joint cup
Specifications
50 mL | |
O-Ring Tap and O-Ring Ball Joint Cup | |
Vacuum/Schlenk Flask | |
Size: 19 |
Product Content Correction
Product Title
GPE Scientific™ J Young Vacuum/Schlenk Flask with O-Ring Tap and O-Ring Ball Joint Cup > Capacity: 50mL
Spot an opportunity for improvement?Share a Content Correction