Learn More
Tocris Bioscience™ JJKK 048
Potent and selective MAGL inhibitor
Brand: Tocris Bioscience™ 5206/10
This item is not returnable.
View return policy
Description
JJKK 048 is a potent and selective monoacylglycerol lipase (MAGL) inhibitor (IC50 = 0.4 nM). Exhibits >13,000 and ∽630-fold selectivity for MAGL over FAAH and ABHD6, respectively. Increases brain levels of 2-arachidonoylglycerol (2-AG) levels in mice in vivo. Promotes analgesia in pain models.
Specifications
>98% | |
JJKK 048 | |
JJKK 048 |
10 mg | |
434.44 |
Product Content Correction
Product Title
Tocris Bioscience™ JJKK 048 > 10mg
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction