missing translation for 'onlineSavingsMsg'
Learn More

Anritsu™ Type K Magnet Probes

Product Code. 11932139
≤400°C, Standard Square, Each
Click to view available options
Temperature (Metric):
≤250°C
≤400°C
Style:
Miniature Round
Standard Square
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11932139

Brand: Anritsu™ MG22KGW1ANP

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Designed with magnets to allow probes to be attached to measurement targets. Anritsu™ Type K Magnet Probes are temperature sensors designed for stationary surfaces.

  • Designed for stationary surface
  • Available with square or round footprint
  • Magnetic probe that attaches to steel surface
  • Thermocouple contact

Specifications

Accuracy ±1.4°C (100°C surface)
Analysis Time 2.0 sec.
Description Head size: 32L x 27W x 34mmD
Temperature (Metric) ≤400°C
Type Thermocouple K
Material Chromel-Alumel
Style Standard Square
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Anritsu™ Type K Magnet Probes >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.