Learn More
Karl Fischer Aqualine™ Water Standard - KF oven , for Karl Fischer titration, Fisher Chemical
Click to view available options
Quantity:
10 g
Specifications
Specifications
Quantity | 10 g |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
59Hours
1Minutes
55Seconds
38New Offer Save Up to 70%
Product Content Correction
Product Title
Karl Fischer Aqualine™ Water Standard - KF oven , for Karl Fischer titration, Fisher Chemical >
Spot an opportunity for improvement?Share a Content Correction