missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCTD1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
360.68 EUR - 575.58 EUR
Specifications
Antigen | KCTD1 |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL |
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18391885
|
Bio-Techne
NBP3-17507-25UL |
25 μg |
360.68 EUR
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18331735
|
Bio-Techne
NBP3-17507-100UL |
100 μg |
575.58 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KCTD1 Polyclonal antibody specifically detects KCTD1 in Human samples. It is validated for ImmunofluorescenceSpecifications
KCTD1 | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
BTB/POZ domain-containing protein KCTD1, C18orf5, potassium channel tetramerisation domain containing 1 | |
This antibody was developed against a recombinant protein corresponding to the amino acids: LGNTYILPKDSQVGPDVKSEAAPKRALYESVFGSGEICGPTSPKRLCIRPSEPVDAVVVVSVKHDPLPLLPEANGHRSTNSPT | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
Polyclonal | |
Purified | |
RUO | |
PBS, pH 7.2, 40% glycerol | |
284252 | |
IgG | |
Affinity purified |