Learn More
Ketanserin tartrate, Tocris Bioscience™
Product Code. 18752664
250 mg
Click to view available options
Quantity:
50 mg
250 mg
Selective 5-HT2A antagonist. Also antagonist at 5-HT1D
Selective 5-HT2A serotonin receptor antagonist; can also be used to discriminate between 5-HT1D and 5-HT1B receptor subtypes.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 83846-83-7
Molecular Formula C26H30FN3O10
Molecular Weight (g/mol) 563.54
MDL Number MFCD00084651,MFCD11864992,MFCD00083392
InChI Key KJJRKCWDAHTVRL-UHFFFAOYNA-N
Synonym ketanserin tartrate
PubChem CID 91885469
IUPAC Name 2,3-dihydroxybutanedioic acid 3-{2-[4-(4-fluorobenzoyl)piperidin-1-yl]ethyl}-1,2,3,4-tetrahydroquinazoline-2,4-dione hydrate
SMILES O.OC(C(O)C(O)=O)C(O)=O.FC1=CC=C(C=C1)C(=O)C1CCN(CCN2C(=O)NC3=CC=CC=C3C2=O)CC1
Specifications
Quantity 250 mg
Formula Weight 545.52
Percent Purity >97%
Chemical Name or Material Ketanserin tartrate
Product Content Correction
Product Title
Ketanserin tartrate, Tocris Bioscience™ >