Learn More
DWK Life Sciences Kimble™ Kontes™ 90mm Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support
Component for microfiltration assembly
Brand: DWK Life Sciences 953752-0090
418.63 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
- Filter size: 90mm
Specifications
Fritted Glass |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
14Hours
7Minutes
24Seconds
15
Product Content Correction
Product Title
DWK Life Sciences Kimble™ Kontes™ 90mm Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support > Base support
Spot an opportunity for improvement?Share a Content Correction