Learn More
DWK Life Sciences Kimble™ Kontes™ Polyethylene Stoppers
Low-cost polyethylene stoppers are an alternative to standard glass and PTFE stoppers.
35.25 EUR - 48.50 EUR
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | ||||||
15327858
|
DWK Life Sciences
774240-0014 |
14/10, 14/20, 14/35 |
35.25 EUR
Pack of 100 |
Estimated Shipment: 07-07-2025
Log in to see stock. |
||||||
15347858
|
DWK Life Sciences
774240-0019 |
19/22 ,19/38 |
40.45 EUR
Pack of 100 |
Estimated Shipment: 07-07-2025
Log in to see stock. |
||||||
15357858
|
DWK Life Sciences
774240-0029 |
29/26, 29/42 |
48.50 EUR
Pack of 100 |
Estimated Shipment: 07-07-2025
Log in to see stock. |
||||||
Description
Low-cost polyethylene stoppers are an alternative to standard glass and PTFE stoppers.
- Leak-proof, air-tight seals
- Wide top flange for ease of use
Specifications
Polyethylene | |
Plug Stopper |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
DWK Life Sciences Kimble™ Kontes™ Polyethylene Stoppers