missing translation for 'onlineSavingsMsg'
Learn More
L-748,337, Tocris Bioscience™ Product Code.: 30001390

L-748,337, Tocris Bioscience™

Product Code. 30001390
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 30001390

Brand: Tocris Bioscience 2760/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective β3 antagonist

Competitive β3-adrenoceptor antagonist that displays selectivity over β1 and β2 receptors (Ki values are 4.0, 204 and 390 nM for β3-, β2- and β1-adrenoceptors respectively). Inhibits cAMP accumulation in response to isoproterenol (IC50 = 6 nM). Reduces iNOS expression, attenuates nitric oxide-induced cell proliferation and induces apoptosis in a melanoma cell line.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 244192-94-7
Molecular Formula C26H31N3O5S
Molecular Weight (g/mol) 497.61
InChI Key AWIONHVPTYTSHZ-DEOSSOPVSA-N
Synonym n-3-2s-2-hydroxy-3-2-4-phenylsulfonyl amino phenyl ethyl amino propoxy phenyl methyl-acetamide, d03vhk, n-3-2s-2-hydroxy-3-2-4-phenylsulfonylamino phenyl ethylamino propoxy phenyl methyl acetamide
PubChem CID 10028830
IUPAC Name N-[[3-[(2S)-3-[2-[4-(benzenesulfonamido)phenyl]ethylamino]-2-hydroxypropoxy]phenyl]methyl]acetamide
SMILES CC(=O)NCC1=CC(=CC=C1)OCC(CNCCC2=CC=C(C=C2)NS(=O)(=O)C3=CC=CC=C3)O

Specifications

Quantity 10 mg
Formula Weight 497.61
Percent Purity >98%
Chemical Name or Material L-748, 337
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-748,337, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.