missing translation for 'onlineSavingsMsg'
Learn More

L 888607 Racemate, MedChemExpress

MedChemExpress L 888607 Racemate is a selective prostaglandin D2 receptor subtype 1 (DP1) antagonist, with Kis of 132 nM and 17 nM for DP1 and thromboxane A2 receptor (TP), respectively.

Encompass_Preferred

446.85 EUR - 3346.65 EUR

Specifications

Chemical Name or Material L 888607 Racemate
CAS 1030017-51-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H15ClFNO2S
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16493066
Encompass_Preferred
View Documents
Medchem Express
HY-111271A
25 mg
1533.60 EUR
25mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16487137
Encompass_Preferred
View Documents
Medchem Express
HY-111271A
5 mg
446.85 EUR
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16485306
Encompass_Preferred
View Documents
Medchem Express
HY-111271A
100 mg
3346.65 EUR
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16454418
Encompass_Preferred
View Documents
Medchem Express
HY-111271A
10 mg
724.95 EUR
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16430737
Encompass_Preferred
View Documents
Medchem Express
HY-111271A
50 mg
2370.60 EUR
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

L 888607 Racemate is a selective prostaglandin D2 receptor subtype 1 (DP1) antagonist, with Kis of 132 nM and 17 nM for DP1 and thromboxane A2 receptor (TP), respectively.
Specifications

Specifications

L 888607 Racemate
Light Yellow
C19H15ClFNO2S
O=C(O)CC1CCN2C1=C(SC3=CC=C(Cl)C=C3)C4=C2C=C(F)C=C4
375.84
Research
Neuroscience-Neuromodulation
1030017-51-6
Research
DMSO : 6 mg/mL (15.96 mM; Need ultrasonic)
375.84
99.44%
Solid
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
L 888607 Racemate, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.