missing translation for 'onlineSavingsMsg'
Learn More
L-Alanine, 99%, MP Biomedicals™ Product Code.: 11442970

L-Alanine, 99%, MP Biomedicals™

Product Code. 11442970 Shop All MP Biomedicals Products
1 kg
Click to view available options
Quantity:
25 g
1 kg
5 kg
This item is not returnable. View return policy

Product Code. 11442970

Brand: MP Biomedicals 0210028701

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A nonessential amino acid that plays a key role in the glucose-alanine cycle between muscle tissue and the liver. In amino acid-degrading tissues such as muscle, amino groups are pooled as glutamate by transamination reactions

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-41-7
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00064410
InChI Key QNAYBMKLOCPYGJ-REOHCLBHSA-N
Synonym l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
PubChem CID 5950
ChEBI CHEBI:16977
IUPAC Name (2S)-2-aminopropanoic acid
SMILES C[C@H](N)C(O)=O

Specifications

Color White
Assay Percent Range 0.99
Density 1.40g/mL
Quantity 1 kg
Formula Weight 89.1
Physical Form Powder
Chemical Name or Material L-Alanine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-Alanine, 99%, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.