missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamine, 99-100%, MP Biomedicals

41.35 EUR - 895.00 EUR

Chemical Identifiers

CAS 56-85-9
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.15
MDL Number MFCD00008044
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
PubChem CID 5961
ChEBI CHEBI:18050
IUPAC Name (2S)-2-amino-4-carbamoylbutanoic acid
SMILES N[C@@H](CCC(N)=O)C(O)=O
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11485192
View Documents
MP Biomedicals
0219467825
25 g
41.35 EUR
25g
Please to purchase this item. Need a web account? Register with us today!
11495192
View Documents
MP Biomedicals
0219467880
100 g
145.00 EUR
100g
Please to purchase this item. Need a web account? Register with us today!
11405202
View Documents
MP Biomedicals
0219467883
250 g
265.00 EUR
250g
Please to purchase this item. Need a web account? Register with us today!
11415202
View Documents
MP Biomedicals
0219467890
500 g
536.00 EUR
500g
Please to purchase this item. Need a web account? Register with us today!
11425202
View Documents
MP Biomedicals
0219467891
1 kg
895.00 EUR
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

56-85-9
146.15
ZDXPYRJPNDTMRX-VKHMYHEASA-N
5961
(2S)-2-amino-4-carbamoylbutanoic acid
C5H10N2O3
MFCD00008044
l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
CHEBI:18050
N[C@@H](CCC(N)=O)C(O)=O

Specifications

56-85-9
MFCD00008044
ZDXPYRJPNDTMRX-VKHMYHEASA-N
(2S)-2-amino-4-carbamoylbutanoic acid
5961
Cell Culture Reagent
L-Glutamine, 99-100%
C5H10N2O3
l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
N[C@@H](CCC(N)=O)C(O)=O
146.15
CHEBI:18050
Crystalline
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room temperature

SDS
Documents

Documents

Product Certifications