missing translation for 'onlineSavingsMsg'
Learn More

L-Glutathione, reduced, 98+%, Thermo Scientific Chemicals

A tripeptide used for reductive cleavage of disulfide bonds

53.37 EUR - 457.28 EUR

Chemical Identifiers

CAS 70-18-8
Molecular Formula C10H17N3O6S
Molecular Weight (g/mol) 307.321
MDL Number MFCD00065939
InChI Key RWSXRVCMGQZWBV-WDSKDSINSA-N
Synonym glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion
PubChem CID 124886
ChEBI CHEBI:16856
IUPAC Name (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid
SMILES C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15484589
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
J62166.06
5 g
53.37 EUR
5g
Please to purchase this item. Need a web account? Register with us today!
15494589
SDS View Documents
Thermo Scientific Alfa Aesar
J62166.14
25 g
138.69 EUR
25g
Please to purchase this item. Need a web account? Register with us today!
15404599
SDS View Documents
Thermo Scientific Alfa Aesar
J62166.22
100 g
457.28 EUR
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

A tripeptide used for the reductive cleavage of disulfide bonds L-Glutathione is used in the treatment of lung diseases for patients who are HIV positive. It protects the cancerous cells by conferring resistance to chemotherapeutic drugs. It is involved in many aspects of metabolism including transport of g-glutanyl amino acids and reductive cleavage of disulfide bonds. As an antioxidant, it prevents damage to important cellular components that arise due to reactive oxygen species like free radicals and peroxide. It is also used to decrease the concentrations of inflammatory cytokines (IL-6, IL-18) as well as involved in increasing the level of serum Ca2+ ions. It is also used in white wine production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, dimethyl formamide, ethanol, phosphate buffered saline, diluted alcohol, liquid ammonia and dimethyl sulfoxide.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 200-725-4

missing translation for 'rtecsNumber' : MC0556000

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-Glutathione, reduced, 98+%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.