missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Leucine, Cell Culture Reagent Product Code.: 15445129

Thermo Scientific Chemicals L-Leucine, Cell Culture Reagent

Product Code. 15445129
500 g
Click to view available options
Quantity:
100 g
250 g
500 g
This item is not returnable. View return policy

Product Code. 15445129

Brand: Thermo Scientific Chemicals J62824.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

In cosmetics industry it is used as antistatic agent, hair conditioning and skin conditioning. L-Leucine acts as a nutrient signal to stimulate protein synthesis. It has also activates the mammalian target of rapamycin kinase that regulates cell growth. Leucine has been used as a molecular marker in the recovery of DNA from paleontological samples for PCR analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In cosmetics industry it is used as antistatic agent, hair conditioning and skin conditioning. L-Leucine acts as a nutrient signal to stimulate protein synthesis. It has also activates the mammalian target of rapamycin kinase that regulates cell growth. Leucine has been used as a molecular marker in the recovery of DNA from paleontological samples for PCR analysis.

Solubility
Soluble in water. Soluble in 1M HCl (100 mg/ml), water (24.3 mg/ml), acetic acid (10.9 mg/ml), and alcohol. Insoluble in ether.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-90-5
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00002617
InChI Key ROHFNLRQFUQHCH-MLHKIVSYNA-N
Synonym l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acid
PubChem CID 6106
ChEBI CHEBI:15603
IUPAC Name (2S)-2-amino-4-methylpentanoic acid
SMILES CC(C)C[C@H](N)C(O)=O

Specifications

Melting Point >300°C
Color White
Beilstein 1721722
Merck Index 14,5451
Solubility Information Soluble in water. Soluble in 1M HCl (100mg/ml),water (24.3mg/ml),acetic acid (10.9mg/ml),and alcohol. Insoluble in ether.
Density 1.293
Quantity 500 g
Formula Weight 131.17
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Leucine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS20K050 Certificate TypeCertificate of Analysis Date29/07/2024 Catalog NumberJ62824.30,J62824.36,J62824.22
Lot NumberU05I003 Certificate TypeCertificate of Analysis Date17/08/2022 Catalog NumberJ62824.30,J62824.36,J62824.22
Lot NumberS02H002 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ62824.30,J62824.36,J62824.22
Lot NumberQ28D022 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ62824.30,J62824.36,J62824.22
Lot NumberW05D020 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ62824.30,J62824.36,J62824.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Leucine, Cell Culture Reagent >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.