missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine Monohydrochloride, Extra Pure, Fisher Chemical™

108.00 EUR - 670.00 EUR

Chemical Identifiers

CAS 657-27-2
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.648
MDL Number 64564
InChI Key BVHLGVCQOALMSV-JEDNCBNOSA-N
Synonym l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisylShow MoreShow Less
PubChem CID 69568
ChEBI CHEBI:53633
IUPAC Name (2S)-2,6-diaminohexanoic acid;hydrochloride
SMILES C(CCN)CC(C(=O)O)N.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10764951
SDS View Documents
Fisher Chemical
L/2700/48
100 g N/A N/A N/A
11478972
SDS View Documents
Fisher Chemical
L/2700/60
1 kg N/A N/A N/A
Description

Description

Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-Lysine Monohydrochloride, Extra Pure, Fisher Chemical™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.