missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Phenylalanine, >99%, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1 kg
Specifications
Specifications
Quantity | 500 g |
Formula Weight | 165.2 |
Grade | Cell Culture |
Chemical Name or Material | L-Phenylalanine, >99% |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
L-Phenylalanine, >99%, MP Biomedicals >
Spot an opportunity for improvement?Share a Content Correction