missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Tryptophanol, 97% Product Code.: 15409038

Thermo Scientific Chemicals L-Tryptophanol, 97%

Product Code. 15409038
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15409038

Brand: Thermo Scientific Chemicals H62506.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-Tryptophanol is an amino acid used in the construction of peptides and proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Tryptophanol is an amino acid used in the construction of peptides and proteins.

Solubility
Soluble in water 11.4 g/L (25°C).

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2899-29-8
Molecular Formula C11H14N2O
Molecular Weight (g/mol) 190.25
MDL Number MFCD00037970
InChI Key UDQCRUSSQAXPJY-UHFFFAOYNA-N
Synonym l-tryptophanol, h-tryptophanol, l---tryptophanol, tryptophanol, s-2-amino-3-1h-indol-3-yl propan-1-ol, 2s-2-amino-3-1h-indol-3-yl propan-1-ol, s-2-amino-3-3-indolyl-1-propanol, s---2-amino-3-3-indolyl propanol, tryptophanol, l, 2-amino-3-1h-indol-3-yl-propan-1-olShow MoreShow Less
PubChem CID 6951149
IUPAC Name 2-amino-3-(1H-indol-3-yl)propan-1-ol
SMILES NC(CO)CC1=CNC2=CC=CC=C12

Specifications

Color Brown
Solubility Information Soluble in water 11.4g/L (25°C).
Quantity 5 g
Formula Weight 190.24
Percent Purity 97%
Physical Form Viscous Liquid
Chemical Name or Material L-Tryptophanol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number81302061 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62506.06,H62506.03
Lot NumberT16E037 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62506.06,H62506.03
Lot NumberZ23F067 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62506.06,H62506.03
Lot NumberX24F015 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62506.06,H62506.03
Lot NumberQ17F021 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62506.06,H62506.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Tryptophanol, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.