missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(-)-Xylose, 99+%

Product Code. 10752651
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-06-3
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00151096
InChI Key PYMYPHUHKUWMLA-UHFFFAOYNA-N
Synonym l-xylose, l +-xylose, l---xylose, 2s,3r,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-xylose, l-xylo-pentose, l-xyl, xylose, l, linear l-xylose, l--xylose
PubChem CID 95259
ChEBI CHEBI:17979
SMILES OCC(O)C(O)C(O)C=O

Specifications

Melting Point 150.0°C to 152.0°C
Color White
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 31,55
Specific Rotation Condition − 18.70 (24.00°C c=4,H2O,24h)
Specific Rotation − 18.70
Solubility Information Solubility in water: soluble. Other solubilities: soluble in pyridine and hot alcohol
IUPAC Name 2,3,4,5-tetrahydroxypentanal
Formula Weight 150.13
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material L(-)-Xylose
Show More Show Less
Product Identifier
  • L(-)-Xylose

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L(-)-Xylose, 99+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.