missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Fast-Freeze™ Flask Adapter

Fast-Freeze™ Flask Adapter for connecting Fast-Freeze™ flasks to freeze-dryer valves.

Brand:  Labconco™ 7545800

 View more versions of this product

Product Code. 10100720

  • 31.05 EUR / Each
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

For connecting Fast-Freeze™ flasks to freeze-dryer valves

  • 40 and 80 mL flasks take size 0.5 in. or 0.5 to 0.75 in.
  • Larger flasks fit size 0.75 in. or large end of 0.5 to 0.75 in.
Specifications

Specifications

Flask Adapter
Borosilicate Glass
For connecting fast freeze™ flasks to freeze-dryer valves
0.75 in.
0.75 in.
4.0 in.
0.5 lb.
40, 80 mL
45° bend
Fast freeze™ flask
1.9 cm
1.905 cm
10.16 cm
0.2 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Labconco™ Fast-Freeze™ Flask Adapter > Material: Glass, Style: 45° Bend, Diameter: 0.75 in.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.