Learn More
Labconco™ SpillStopper™ Work Surface for 6ft. Protector™XStream
Labconco SpillStopper™ Work Surface is dished and contoured to conform to the interior liner to contain spills.
Brand:  Labconco™ 9503610
 View more versions of this product
Product Code. 13141144
Description
Description
  • Dished and contoured to conform to interior liner of Protector Laboratory Hoods
  • Front edge with large radius to aerodynamically direct airflow in hood
  • Pre-drilled holes for venting and plumbing passthroughs
  • Includes left rear cupsink cutout
Specifications
Specifications
Protector XStream Fume Hood (72 in.)
Work Surface
Laboratory Fume Hood Work
Left Rear
Epoxy Resin
Documents
One moment while we fetch your results.
Product Title
Labconco™ SpillStopper™ Work Surface for 6ft. Protector™XStream > Left rear