Learn More
Buerkle™ LaboPlast™ Disposable Scoop
Disposable scoop made of polystyrene is produced and assembled in a cleanroom class 7 and individually packaged for disposable use. The disposable scoop is ideal for powders, granulates, and pastes.
Brand: Buerkle™ 5378-0008
This item is not returnable.
View return policy
Description
Scoop, disposable with or without cover
- Polystyrene, white
- Individually packaged
- Production and packaging according to Cleanroom Class 7
- With EU Foodstuffs and FDA approval
Specifications
Disposable Scoop | |
With Cover | |
150 mL | |
218 mm |
Polystyrene | |
With Cover | |
Single-use |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Videos
-
Buerkle Sample scoops SteriPlast and LaboPlast
Brochure LaboPlast SteriPlast scoops spoons spatulas
File size: 608.8 kB
Topsellers disposamplers
File size: 1.3 MB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
244824 | Certificate of Analysis | 04/12/2024 | 5378-0008 |
235456 | Certificate of Compliance | 19/12/2023 | 5378-0008 |
235296 | Certificate of Compliance | 05/12/2023 | 5378-0008 |
223909 | Certificate of Compliance | 06/10/2022 | 5378-0008 |
222264 | Certificate of Compliance | 02/06/2022 | 5378-0008 |
Product Content Correction
Product Title
Buerkle™ LaboPlast™ Disposable Scoop > 150 mL; with cover
Spot an opportunity for improvement?Share a Content Correction