Learn More
Buerkle™ LaboPlast™ Disposable Scoop
Disposable scoop made of polystyrene is produced and assembled in a cleanroom class 7 and individually packaged for disposable use. The disposable scoop is ideal for powders, granulates, and pastes.
Brand: Buerkle™ 5378-0002
This item is not returnable.
View return policy
Description
Scoop, disposable with or without cover
- Polystyrene, white
- Individually packaged
- Production and packaging according to Cleanroom Class 7
- With EU Foodstuffs and FDA approval
Specifications
Disposable Scoop | |
With Cover | |
25 mL | |
142 mm |
Polystyrene | |
With Cover | |
Single-use |
Videos
-
Buerkle Sample scoops SteriPlast and LaboPlast
Brochure LaboPlast SteriPlast scoops spoons spatulas
File size: 608.8 kB
Topsellers disposamplers
File size: 1.3 MB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Product Content Correction
Product Title
Buerkle™ LaboPlast™ Disposable Scoop > 25 mL; with cover
Spot an opportunity for improvement?Share a Content Correction