Learn More
Buerkle™ LaboPlast™ Disposable Scoop
BIOPHARMAGREENER_CHOICE
Disposable scoop made of polystyrene is produced and assembled in a cleanroom class 7 and individually packaged for disposable use. The disposable scoop is ideal for powders, granulates, and pastes.
Brand:  Buerkle™ 5378-0006
 View more versions of this product
Product Code. 15396307
  • 36.35 EUR / Pack of 10
Estimated Shipment: 09-06-2025
Description
Scoop, disposable with or without cover
  • Polystyrene, white
  • Individually packaged
  • Production and packaging according to Cleanroom Class 7
  • With EU Foodstuffs and FDA approval
Specifications
Specifications
Disposable Scoop
With Cover
100 mL
206 mm
Polystyrene
With Cover
Single-use
Videos
  • Buerkle - Sample scoops SteriPlast and LaboPlast
    The disposable sample scoop is ideal for powders, granulates and pastes.
    Video Player is loading.
    Current Time 0:00
    Duration 1:10
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 1:10
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Buerkle Sample scoops SteriPlast and LaboPlast
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number141219BW53780 Certificate TypeCertificate of Compliance Date29/03/2023 Catalog Number5378-0006
Lot Number231299 Certificate TypeCertificate of Compliance Date28/03/2023 Catalog Number5378-0006
Product Title
Buerkle™ LaboPlast™ Disposable Scoop > 100 mL; with cover