Learn More
Epredia™ Laboratory Marking Pen, Secureline Marker II
Mark cassettes and microscope slides with the Epredia™ Laboratory Marking Pen.
16.80 EUR - 51.75 EUR
Specifications
Color Black
View More Specs
Products 2
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
15582321
View Documents
Epredia™
2000
Laboratory Marking Pen N/A N/A N/A
12657736
View Documents
Epredia™
1451
Secureline Marker II N/A N/A N/A
Description
Description
  • Resistant to common laboratory solvents (water, 10% neutral buffered formalin, alcohol)
  • Extra fine point for precise writing
  • Non-sterile
  • Not made with natural rubber latex
Specifications
Specifications
Black
Documents
Product Certifications
Product Content Correction
Product Title
Epredia™ Laboratory Marking Pen, Secureline Marker II