missing translation for 'onlineSavingsMsg'
Learn More

Lambda Light Chain Ab-1 Mouse Monoclonal Antibody, Epredia™

Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ Lambda Light Chain Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia MS-180-P1ABX

 View more versions of this product

Product Code. 12660167

This item is not returnable. View return policy

Description

Description

Antibody to the λ light chain of immunoglobulin is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Mouse

Clone: HP6054

Isotype: IgG2a/k

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Not determined

Immunogen: Purified human IgG myeloma proteins covalently coupled to polyaminostyrene micro beads

Molecular Weight: 25kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Lambda Light Chain Ab-1
Monoclonal
1mg/mL
Mouse
100 μL
Hematopoietic
Human
IgG2a κ
Immunohistochemistry (Paraffin)
HP6054
Unconjugated
Purified human IgG myeloma proteins covalently coupled to polyaminostyrene micro beads
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.