Learn More
Lambda Light Chain Ab-1 Mouse Monoclonal Antibody, Epredia™
Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ Lambda Light Chain Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
129.54 EUR - 520.20 EUR
Specifications
Antigen | Lambda Light Chain Ab-1 |
---|---|
Clone | HP6054 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12650167
|
Epredia
MS-180-P0 |
Concentrated | 100 μL | N/A | |||||
12660167
|
Epredia
MS-180-P1ABX |
Concentrated | 100 μL | N/A | |||||
12690167
|
Epredia
MS-180-PABX |
Concentrated | 200 μL | N/A | |||||
12670167
|
Epredia
MS-180-P1 |
Concentrated | 500 μL | N/A | |||||
12620167
|
Epredia
MS-180-P |
Concentrated | 1 mL |
372.00 EUR
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12610177
|
Epredia
MS-180-R7 |
Prediluted | 7 mL |
216.00 EUR
7mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Antibody to the λ light chain of immunoglobulin is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.Host Species: Mouse
Clone: HP6054
Isotype: IgG2a/k
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: Not determined
Immunogen: Purified human IgG myeloma proteins covalently coupled to polyaminostyrene micro beads
Molecular Weight: 25kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Lambda Light Chain Ab-1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Human | |
IgG2a κ |
HP6054 | |
Monoclonal | |
Mouse | |
Hematopoietic | |
Purified human IgG myeloma proteins covalently coupled to polyaminostyrene micro beads | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.