Learn More
Thermo Scientific™ LanthaScreen™ Tb-p-c-jun (pSer73) Antibody
For use in combination with the GFP-c-Jun physiological protein substrate (PV4446) to evaluate inhibitors in medium to high throughput screening applications.
Brand: Thermo Scientific™ PV4449
This item is not returnable.
View return policy
Description
- A part of the activating protein-1 (AP-1) transcription factor, c-Jun is phosphorylated at Ser73 by members of the JNK family
- For use in combination with the GFP-c-Jun physiological protein substrate (PV4446) to evaluate inhibitors in medium to high throughput screening applications
Specifications
Tb (Terbium)/Lanthanide Chelates | |
Terbium-labeled p-c-Jun (pSer73) antibody in HEPES buffered saline. Store at -20°C. | |
Human | |
End Point | |
Ser/Thr Kinases |
25 μg | |
340/490, 520 | |
Dry Ice | |
Hu | |
LanthaScreen™ |
Product Content Correction
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction