missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ Tb-p-c-jun (pSer73) Antibody
For use in combination with the GFP-c-Jun physiological protein substrate (PV4446) to evaluate inhibitors in medium to high throughput screening applications.
4215.00 EUR - 20450.00 EUR
Specifications
Conjugate | Tb (Terbium)/Lanthanide Chelates |
---|---|
Excitation/Emission | 340/490, 520 |
Product Line | LanthaScreen™ |
Reactivity | Human |
Readout | End Point |
Description
- A part of the activating protein-1 (AP-1) transcription factor, c-Jun is phosphorylated at Ser73 by members of the JNK family
- For use in combination with the GFP-c-Jun physiological protein substrate (PV4446) to evaluate inhibitors in medium to high throughput screening applications
Specifications
Tb (Terbium)/Lanthanide Chelates | |
LanthaScreen™ | |
End Point | |
Ser/Thr Kinases |
340/490, 520 | |
Human | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title