Learn More
Thermo Scientific™ Legionella BMPA Selective Agar
Isolate Legionella from clinical specimens with a selective microbiological medium which yields optimal recovery of Legionellaceae in short incubation periods.
38.85 EUR
Specifications
Format | 90mm Monoplate |
---|---|
Media Type | Legionella BMPA Selective Agar |
Product Type | Agar |
Packaging Type | Flow Wrapped - Single Layer |
Quantity | 10/Pk. |
Product Code | Brand | Quantity | Format | Media Type | Manufacturing Origin | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Format | Media Type | Manufacturing Origin | Price | Quantity & Availability | |||
13285349
|
Thermo Scientific™
PO5035A |
10/Pk. | 90mm Monoplate | Legionella BMPA Selective Agar | Germany |
38.85 EUR
Pack of 10 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
|||
Description
Isolate Legionella from clinical specimens using Thermo Scientific™ Legionella BMPA Selective Agar, a selective medium that exhibits optimal recovery of Legionellaceae in short incubation periods from both environmental samples and clinical specimens.
Legionella are aerobic, motile, nutritionally fastidious, pleomorphic, gram-negative rods that require special growth factors in culture media to grow. Transmission occurs through aerosolization or aspiration of contaminated water, not person-to-person. Legionellosis refers to two distinct clinical syndromes: Legionnaires’ disease, which most often presents as severe pneumonia, and Pontiac fever, which is an acute, febrile, self-limiting, viral-like illness. Over forty species of Legionella have been characterized; however, Legionella pneumophila remains the most prevalent etiological agent of Legionnaires’ disease. Legionella can be isolated from respiratory tract secretions of all types, sterile body fluids such as blood, and biopsy material.This product may not be available in all countries or territories. Please consult your local representative for availability.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
90mm Monoplate | |
Agar | |
10/Pk. |
Legionella BMPA Selective Agar | |
Flow Wrapped - Single Layer |
Safety and Handling
missing translation for 'shelfLife' : 8 Weeks
Your input is important to us. Please complete this form to provide feedback related to the content on this product.