missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) Rapid Atto565 Antibody Labeling Kit

Applications: Flow Cytometry

Brand:  Novus Biologicals™ 351-0010

 View more versions of this product

Product Code. 18728113

  • 699.00 EUR / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Atto565 is one of a new generation of fluorescent labels. It has a strong absorption at 563nm, high fluorescence at 592nm (extinction coefficient 1.2 x10&superscript_begin;5&superscript_end; cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;) and quantum yield.

Order Info

Quantity

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at -20°C. Avoid freeze-thaw cycles.
300 μg
Atto 565
Rapid Atto565 Antibody Labeling Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novus Biologicals™ Lightning-Link (R) Rapid Atto565 Antibody Labeling Kit > 3 x 200μg Reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.