Learn More
Lithium acetate dihydrate, 99%, Thermo Scientific Chemicals
Description
Lithium acetate dihydrate is used as a buffer component in the modified ninhydrin reagent for amino analysis by ion-exchange chromatography. It is used as a catalyst in organic reactions, alkyd resin and acrylic polymer production and as an anticorrosion agent in molding polyphenylene sulfide resins as well as in the separation of saturated and unsaturated fatty acids. In the pharmaceutical industry, it is utilized for the preparation of diuretics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lithium acetate dihydrate is used as a buffer component in the modified ninhydrin reagent for amino analysis by ion-exchange chromatography. It is used as a catalyst in organic reactions, alkyd resin and acrylic polymer production and as an anticorrosion agent in molding polyphenylene sulfide resins as well as in the separation of saturated and unsaturated fatty acids. In the pharmaceutical industry, it is utilized for the preparation of diuretics.
Solubility
Soluble in water and ethanol.
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 54°C to 57°C |
Density | 1.3 |
Quantity | 1000 g |
Beilstein | 3564320 |
Merck Index | 14,5521 |
Solubility Information | Soluble in water and ethanol. |
Formula Weight | 102.01 (65.98 Anhydrous) |
Percent Purity | 99% |
Chemical Name or Material | Lithium acetate dihydrate |
Safety and Handling

- Lithium acetate dihydrate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Lithium acetate dihydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.