missing translation for 'onlineSavingsMsg'
Learn More
Lithium sulfate hydrate, Puratronic™, 99.996% (metals basis), Thermo Scientific Chemicals Product Code.: 11362078

Lithium sulfate hydrate, Puratronic™, 99.996% (metals basis), Thermo Scientific Chemicals

Product Code. 11362078
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11362078

Brand: Thermo Scientific Alfa Aesar 012983.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10377-48-7
Molecular Formula Li2O4S
Molecular Weight (g/mol) 109.94
MDL Number MFCD00011086
InChI Key INHCSSUBVCNVSK-UHFFFAOYSA-L
Synonym lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrous
PubChem CID 66320
ChEBI CHEBI:53474
SMILES [Li+].[Li+].[O-]S([O-])(=O)=O

Specifications

Melting Point 130°C (in -H2O)
Color White
Physical Form Crystalline
Quantity 25 g
Linear Formula Li2SO4·xH2O
Merck Index 14,5541
IUPAC Name dilithium(1+) sulfate
Formula Weight 109.94 (Anhydrous)
Percent Purity 99.996%
Grade Puratronic™
Density 2.06 g/mL
Assay (metals basis)
Chemical Name or Material Lithium sulfate hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lithium sulfate hydrate, Puratronic™, 99.996% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.