Learn More
Brannan™ LO-tox™ Lab Thermometer with 3-points UKAS Calibration
Click to view available options
Temperature Range (Metric):
-1°C to 101°C
-1°C to 51°C
-10°C to +110°C
-10°C to +210°C
-10°C to +260°C
-10°C to +300°C
-10°C to +360°C
-10°C to +50°C
-20°C to +110°C
-20°C to +150°C
-35°C to +50°C
Subdivision:
0.1°C
0.2°C
0.5°C
1°C
2°C
Immersion Type:
Partial Immersion
Total Immersion
Length (Metric):
15.5 cm
30.5 cm
40.5 cm
Förpackningsstorlek
Each
Beskrivelse
- LO-tox™ is an organic filling specially chosen to achieve a wide temperature span and provide the lowest practical levels of hazard
- Safe non-mercury filling
- Supplied with calibration certificate
- Non-roll end cap
- Supplied as standard in protective individual, resealable telescopic tube
- Made in UK
Tekniske data
Tekniske data
| Calibration Points | 0°C, 100°C, 250°C |
| Certifications/Compliance | ISO 17025 UKAS |
| Immersion Type | Partial Immersion |
| Length (Metric) | 30.5 cm |
| Material | Glass |
| Thermometer Fill | Brown Spirit |
| Max. Temperature (English) | 680°F |
| Max. Temperature (Metric) | +360°C |
| Min. Temperature (English) | 14°F |
| Min. Temperature (Metric) | -10°C |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion