Learn More
Lonza™ AccuGENE™ 1M Tris HCl Buffers
Ready-to-use solutions ideal for a wide range of molecular biology applications
Brand:  Lonza™ 51236
 View more versions of this product
Product Code. 11552096
  • 151.00 EUR / 1 L
Explore more special offers
Description
Description
  • Prepared with 18 megohm water
  • Filtered using 0.2μ filter and filled into sterile bottles
  • Manufactured according to strict quality control standards to ensure consistency
  • Guaranteed DNase, RNase and protease-free
  • Customized solutions are available to meet individual needs
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 M
Ideal for a wide range of molecular biology applications
1000 mL
Yes
7.2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For research or manufacturing use only.