missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ AccuGENE™ 1M Tris HCl Buffers

Ready-to-use solutions ideal for a wide range of molecular biology applications

123.00 EUR - 151.00 EUR

Specifications

Concentration 1 M
DNase Yes
For Use With (Application) Ideal for a wide range of molecular biology applications
Quantity 1000 mL
View More Specs

Products 3
Product Code Brand pH Price Quantity & Availability  
Product Code Brand pH Price Quantity & Availability  
11562096
View Documents
Lonza™
51237
7.4
123.00 EUR
Each
Please to purchase this item. Need a web account? Register with us today!
11552096
View Documents
Lonza™
51236
7.2
151.00 EUR
1 L
Please to purchase this item. Need a web account? Register with us today!
11572096
View Documents
Lonza™
51238
8.0
123.00 EUR
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Prepared with 18 megohm water
  • Filtered using 0.2μ filter and filled into sterile bottles
  • Manufactured according to strict quality control standards to ensure consistency
  • Guaranteed DNase, RNase and protease-free
  • Customized solutions are available to meet individual needs

Specifications

Specifications

1 M
Yes
Ideal for a wide range of molecular biology applications
1000 mL
Videos
SDS
Documents

Documents

Product Certifications

For research or manufacturing use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.