missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ BioWhittaker™ Dulbecco's Modified Eagle's Medium with 1.0g L-Glucose per Liter

DMEM is used in a wide range of mammalian cell culture. BioWhittaker™ Dulbecco's Modified Eagle's Medium with 1.0g L-Glucose per Liter is manufactured in accordance with cGMP regulations.

Brand:  Lonza™ BE12-707F

 View more versions of this product

Product Code. 11635220

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • DMEM is a modification of Basal Medium Eagle (BME) that contains four-times the concentrations of the amino acids and vitamins.
  • Suitable for most types of cells, including human, monkey, hamster, rat, mouse, chicken and fish cells. All Manufactured in accordance with cGMP regulations
  • The formulations used for standard liquid media products are those recommended by the Tissue Culture Association
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2° to 8°C
DMEM Media
Sterile-filtered through two consecutive 0.2 μm filters
No Glutamine
500 mL
Liquid
Sterile
Glucose
7 to 7.4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.