missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ DNA QuantLadders
Provide convenient, visual quantitation of DNA within ±1% accuracy
Brand: Lonza™ 50335
This item is not returnable.
View return policy
Description
• Ladders are prepared from five highly purified DNA fragments in perfect relative mass equivalence
• Reverse QuantLadders reverse the mass equivalence order of the QuantLadder
• SimplyLoad™ ready-to-load ladders contain loading buffer; other ladders are supplied with separate loading buffer
• Store at 2° to 8°C for up to 12 months
Specifications
DNA Reverse QuantLadder | |
100 μg/mL | |
Liquid | |
Quantitation of DNA within ±1% accuracy | |
125 μL |
TE | |
DNA Molecular Weight Marker/Ladder | |
Includes a vial of 6X sample loading buffer 1mL total volume 15% w/v Ficoll™ 400, 0.06% w/v Bromophenol Blue, 0.06% w/v Xylene Cyanol FF 30mM EDTA | |
5 | |
2° to 8°C |